Learn More
Invitrogen™ Canine Parvovirus Monoclonal Antibody (H69D)

Description
MA1-7014 detects the canine Parvovirus from infected samples. MA1-7014 has been successfully used in ELISA procedures. The MA1-7014 immunogen is canine Parvovirus lysate.
Specifications
Specifications
| Antigen | Canine Parvovirus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | H69D |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CPV; parvo |
| Host Species | Mouse |
| Immunogen | Canine Parvovirus lysate. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.