Learn More
Kingfisher Biotech Inc Canine PD-L1 ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY1414D004
Encompass_Preferred
The Canine PD-L1 ELISA contains capture antibody standard and detection antibody for development of a Canine PD-L1 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Canine PD-L1 ELISA can also be used to measure Dingo PD-L1. Alternate names include CD274 B7-H B7H1 PD-L1 PDCD1L1 PDCD1LG1 PDL1 CD274 molecule Programmed cell death 1 ligand 1. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5448
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.