Learn More
Kingfisher Biotech Inc Canine PDGF-BB Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1322D005
Encompass
The Canine PDGF-BB yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine PDGF-BB applications are for cell culture, ELISA standard, and Western Blot Control. Canine PDGF-BB yeast-derived recombinant protein can be purchased in multiple sizes. Canine PDGF-BB Specifications: (Molecular Weight: 12.3 kDa) (Amino Acid Sequence: SLGSPTVAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPTFKKATVT LEDHLACKCE TVVAARPVT (109)) (Gene ID: 442986). For research use only. Made in the USA
Catalog No. 50-253-3396
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.