Learn More
Kingfisher Biotech Inc Canine TNF alpha Biotinylated Recombinant Protein
Supplier: Kingfisher Biotech Inc RPB1861D010
The Canine TNF alpha Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine TNF alpha Biotinylated applications are for cell culture. Canine TNF alpha Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Canine TNF alpha Biotinylated Specifications: (Molecular Weight: 17.3 kDa) (Amino Acid Sequence: VKSSSRTPSD KPVAHVVANP EAEGQLQWLS RRANALLANG VELTDNQLIV PSDGLYLIYS QVLFKGQGCP STHVLLTHTI SRFAVSYQTK VNLLSAIKSP CQRETPEGTE AKPWYEPIYL GGVFQLEKGD RLSAEINLPN YLDFAESGQV YFGIIAL (157)) (Gene ID: 403922). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.