Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine TNF-alpha Recombinant Protein

Catalog No. ENRCTNFAI
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRCTNFAI 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRCTNFAI Supplier Invitrogen™ Supplier No. RCTNFAI
Add to Cart
Add to Cart

Recombinant Protein

For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: The biological activity of recombinant canine TNFa was measured by its cytolytic effect on mouse WEHI-13VAR cells in the presence of Actinomycin D. The ED50 (or 1 unit of activity) for this effect in our laboratories is typically 10 pg/mL. Refs: Kitani, H. et al. (2002) Infect. Immun. 70:2210. Khabar, K.S. et al. (1995) Immunol. Lett. 46:107. Expression: The mature, soluble form of canine TNF-alpha was expressed in Pichia pastoris, X-33. GenBank™ Accession # P51742, amino acids 77 - 233. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: VKSSSRTPSD. Reconstitute with 0.5 mL of distilled water.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51742
For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 403922
Molecular Weight (g/mol) 17.3 kDa
Name Canine TNF-alpha
Quantity 20 μg
Source Yeast
Regulatory Status RUO
Gene Alias APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1 F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Common Name TNF alpha
Gene Symbol TNF
Biological Activity ED50 = 10 pg/mL; determined by the dose-dependent cytolytic effect on mouse WEHI-13 VAR cells in the presence of Actinomycin D.
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Sequence Canine TNF-alpha, amino acids 77-233 (Accession # P51742)
Content And Storage 4°C
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.