Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cannabinoid Receptor 1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA1745
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1745 Supplier Thermo Scientific Supplier No. PA1745
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-745 detects cannabinoid receptor 1 (CB1) from human, rat, monkey, mouse, chicken, salamander and goldfish tissues. This antibody detects a few non-specific bands in NIH-3T3 and HT-29 cell lysates. PA1-745 has been successfully used in Western blot, FACS, immunohistochemistry, and immunocytochemistry procedures. By Western blot, this antibody detects a ∽60 kDa protein representing CB1 and less intense bands of ∽23 kDa, ∽72 kDa, and ∽180 kDa from rat brain homogenate. Immunocytochemical staining of CB1 in living or fixed cultured hippocampal neurons with PA1-745 results in intense, punctate staining along processes, with little staining over cell bodies. PA1-745 antigen is a fusion protein containing the first 77 amino acid residues of the rat CB1 receptor.

This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cannabinoid Receptor 1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CNR1
Gene Accession No. P20272, P21554, P47746
Gene Alias brain-type cannabinoid receptor, CANN6, Cannabinoid receptor 1, cannabinoid receptor 1 (brain), CB1, CB1 cannabinoid receptor, CB1 receptor, CB1A, CB1K5, CB1R, CB-R, central cannabinoid receptor, CNR, Cnr1, HGNC:2159, OTTHUMP00000016839, RP1-23D17.1, seven transmembrane domains, G-protein-coupled receptor, Skr6, SKR6R, striatal cannabinoid receptor type 1 protein
Gene Symbols CNR1
Host Species Rabbit
Immunogen A fusion protein containing the first 77 amino acid residues from rat CB1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1268, 12801, 25248, 428633
Target Species Amphibian, Chicken, Fish, Human, Mouse
Content And Storage -20°, C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.