Learn More
Apexbio Technology LLC Sennoside B 128-57-4 20mg
SDP
Supplier:  Apexbio Technology LLC N167720
Encompass_Preferred
Sennoside B (CAS 128-57-4) is a naturally occurring anthraquinone glycoside present in various organisms including puffer fish salamanders and amphibian skin As a known stimulant laxative Sennoside B acts primarily by increasing colonic peristalsis through irritation of the intestinal mucosa facilitating bowel movements This mechanism involves the hydrolysis of sennosides by intestinal bacteria resulting in active metabolites that modulate water and electrolyte secretion in the colon In biomedical research Sennoside B is utilized for studies on gastrointestinal motility drug transport and as a model compound in assays assessing intestinal drug absorption and excretion dynamics
Catalog No. 50-000-87722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.