Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cap for 15 mL Centrifuge tube

Catalog No. 07000513 Shop All Greiner Bio-One Products
Change view
Click to view available options
Product Type:
Centrifuge Tube Cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
07-000-513 Centrifuge Tube Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-513 Supplier Greiner Bio-One Supplier No. 315374

missing translation for 'validMainframeMsg'

For use with 15mL centrifuge tube

  • For use with 15mL centrifuge tube
TRUSTED_SUSTAINABILITY

Specifications

Product Type Centrifuge Tube Cap
For Use With (Equipment) 15 mL Centrifuge Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.