Learn More
Microsolv Technology Corporation Cap For Screw Top Vials 10-425
Supplier:  Microsolv Technology Corporation 9503S0CK
Encompass_Preferred
Caps, Screw Top, with an Open Hole. knurled, polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 10-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.