Learn More
McKesson General Medical CAP-LL TIP RED ST
Supplier:  McKesson General Medical 388213
Encompass
Luer lock tip cap for standard luer lock syringes. Packaged individually. Beautiful red color. Manufacturer Part Number: H93865001.
Catalog No. NC1274556