Learn More
Thomas Scientific Racked Tubes - 2.1ml, Mat of 96 caps, perforated for strips of 8, Qty: 10
SDP
Supplier:  Thomas Scientific 1159N35
Encompass
Standard 96 well format These tubes use all the space in the 8 x 12 format, facilitating multi-step dilutions and preps, without transferring samples from larger tubes. Compatible with multi-channel harvesters, pipettes, Biomek, and many rotors. Tubes are translucent polypropylene.
Catalog No. 50-145-8242
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.