Learn More
MilliporeSigma™ Supelco™ Cap for Mobile Phase Reservoir System, Bottle Cap GL 45
Catalog No. 111005155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-5155 1/Pk.
1 options
Catalog No. 11-100-5155 Supplier MilliporeSigma™ Supelco™ Supplier No. 55062
Add to Cart
Add to Cart
Cap fits 1-liter or 2-liter reservoir system bottles. Cap insert has two holes for 1/8 in. tubing, one for luer taper
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Packaging Quantity 1/Pk.
Description Bottle cap GL 45; Number of holes 3
Material Polypropylene
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company