Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cap Strip for 8-Tube Real Time PCR Tube Strip

Catalog No. 07000535 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-535 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-535 Supplier Greiner Bio-One Supplier No. 373250
Add to Cart
Add to Cart

For use with PCR 8-tube Strip. Greiner Bio-One Cap Strip for 8-Tube PCR Tube Strip is thin walled for optimal heat transfer.

  • Non-Pyrogenic
  • Flat lid
  • Polypropylene
  • Free of detectable DNase, RNase, human DNA
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
  • Seals 8-tube strips in addition to Greiner Bio-One poly
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Description Compatible with most Real Time PCR devices, such as ABI Prism 7700, MX4000 (Stratagene) oder DNA Engine Opticon (MJ Research)
Quantity 1250/Cs.
For Use With (Equipment) PCR 8-tube Strip
Sterility Non-sterile
Material Polypropylene
Product Type Cap Strip

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.