Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cap Strips for PCR 8-tube Strips

Catalog No. 07000536 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-536 Red
07-000-537 Blue
07-000-538 Green
07-000-539 Yellow
07-000-540 Violet
07-000-541 Assorted
07-000-309 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 07-000-536 Supplier Greiner Bio-One Supplier No. 373273
Add to Cart
Add to Cart

For use with PCR 8-tube Strip. Greiner Bio-One Cap Strips are thin walled for optimal heat transfer.

  • Non-Pyrogenic
  • Domed lid
  • Highly transparent polypropylene
  • Free of detectable DNase, RNase, human DNA
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
  • Seals 8-tube strips in addition to
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Quantity 1250/Cs.
For Use With (Equipment) PCR 8-tube Strip
Sterility Non-sterile
Material Polypropylene
Product Type Cap Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.