Learn More
Enzo Life Sciences Caspase-8 (mouse) monoclonal antibody (1G12) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX804447C100
Encompass_Preferred
Clone: 1G12. Host: Rat. Isotype: IgG1. Immunogen: Recombinant mouse caspase-8 p18 subunit. UniProt: O89110. Species Reactivity: Mouse. Specificity: Recognizes the p18 subunit of caspase-8. Crossreactivity: Does not cross-react with human caspase-8. Applications: ELISA. Flow Cytometry, ICC, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide and 50% glycerol. Handling: For. Long Term Storage, aliquot and freeze at -20°C. Avoid freeze/thaw cycles.
Catalog No. NC9938025