Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capecitabine, 99%

Catalog No. ac456241000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456241000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456241000 Supplier Thermo Scientific Chemicals Supplier No. 456241000
Only null left
Add to Cart

CAS: 154361-50-9 | C15H22FN3O6 | 359.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154361-50-9
Molecular Formula C15H22FN3O6
Molecular Weight (g/mol) 359.35
InChI Key GAGWJHPBXLXJQN-UORFTKCHSA-N
Synonym capecitabine, xeloda, capiibine, capecitibine, caxeta, xabine, capecitabine usan, capecitabina, capecitabinum, capecitabin
PubChem CID 60953
ChEBI CHEBI:31348
IUPAC Name pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
SMILES CCCCCOC(=O)NC1=NC(=O)N(C=C1F)C2C(C(C(O2)C)O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Formula Weight 359.35
Percent Purity 99%
Chemical Name or Material Capecitabine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May damage fertility.
May damage the unborn child.
May cause cancer.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.