Learn More
Molex Molex-Polymicro Capillary 10UMx150UM 10M Reel
Supplier:  Molex 1068150004/10M
Encompass_Preferred
TSP010150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 150um OD, spool length of 10m.
Catalog No. 50-110-8570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.