Learn More
Molex Molex-Polymicro Capillary 50UMx363UM 10M Reel
Supplier:  Molex 1068150017/10M
Encompass_Preferred
TSP050375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 50um ID x 363um OD, spool length of 10m.
Catalog No. 50-110-7680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.