Learn More
Molex Molex-Polymicro Capillary 75UMx150UM 50M Reel
Supplier: Molex 1068150018/50M
TSP075150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 150um OD, spool length of 50m.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.