Learn More
AB Sciex LLC Bare Fused-Silica Capillary, 75 µm ID X 50 cm
Supplier:  AB Sciex LLC 338454
Encompass
Capillary for use in PA 800 series and P/ACE MDQ Capillary Cartridges for separation and analysis of a variety of samples including pharmaceuticals industrial chemicals chiral drugs nucleic acids amino acids proteins and peptides. Capillaries must be trimmed to optimize speed/resolution.
Catalog No. NC1002497
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.