Learn More
Dynalon™ Glass Capillary Tubes with One End Sealed
SCI_ED
Catalog No. s37565 Shop All Dynalab Corporation Products
Change view
Click to view available options
For Use With (Equipment):
DMP100 Melting Point Apparatus
Catalog No. For Use With (Equipment)
S37565 DMP100 Melting Point Apparatus
1 options
Catalog No. S37565 Supplier Dynalon™ Supplier No. DMP100/10
Only null left
Add to Cart
Edge
Add to Cart
Economical glass capillary tubes are for use with DMP100 Melting Point Device.
  • Transparent glass tube
  • Packaged in a tube of 100 each
  • Length: 100mm
  • Determine chemical melting point and purity of solid chemical by adding 4 to 6mm to the tube and inserting it into the Melting Point Device
Specifications
Product Type Glass Capillary Tubes
For Use With (Equipment) DMP100 Melting Point Apparatus