Learn More
3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness
Catalog No. 19809573 Shop All 3M Products
Change view
Click to view available options
3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness,Medium
Catalog No.
19-809-573 3M™ DBI-SALA™ ExoFit NEX™ Tower Climbing Harness,Medium
1 options
Catalog No. 19-809-573 Supplier 3M™ Supplier No. 7100188765/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Aluminum back and front D-rings, belt with pad and side D-rings, seat sling with suspension D-rings, locking quick connect buckle leg straps, comfort padding.
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite™ aluminum back D-ring
  • Tech-Lite™ aluminum front D-ring
  • Body belt/hip pad with Tech-Lite™ aluminum side D-rings
  • Removable seat sling with aluminum board and suspension D-rings
  • Duo-Lok™ quick connect chest and leg straps
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 9.7 lb.
D-Ring Location Back, Side, Front, Suspension
Product Type Tower Climbing Harness
Size Medium
Webbing Material Repel™ Polyester
Certifications/Compliance ANSI A10.32, ANSI Z359.1, ANSI Z359.4, OSHA 1926.502, ANSI Z359.3, OSHA 1910.66