Learn More
3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Harness
Catalog No. 19810564 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-564 X-Large
19-810-561 Small
19-810-562 Medium
19-810-563 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-810-564 Supplier 3M™ Supplier No. 7100237065/DEL
Aluminum back D-ring, locking quick connect buckle leg straps, comfort padding.
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite™ aluminum back D-ring
  • Duo-Lok™ quick connect chest and leg straps
  • Repel™ webbing
  • Protective shoulder caps
  • Loops for body belt
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Repel Polyester
Weight (English) 4 lb.
Certifications/Compliance ANSI Z359.4, ANSI Z359.1, OSHA 1910.66, ANSI A10.32, OSHA 1926.502
Webbing Material Repel™ Polyester
D-Ring Location Back
Product Type Vest-Style Harness
Size X-Large
Tensile Strength (English) 6000 lb.