Learn More
3M™ DBI-Sala™ ExoFit™ XP Vest-Style Full Body Harness
Catalog No. 19169851 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
19-169-851 Medium
1 options
Catalog No. 19-169-851 Supplier 3M™ Supplier No. 1110101
Designed based on the concept of single piece of material constructed in shape of X that wraps around for the ultimate in no-tangle donning and comfortable security
  • Unique spring loaded stand-up dorsal D-ring ensures fast, easy and safe connections
  • Ergonomic design incorporates built-in shoulder, hip and leg padding
  • Soft edging moves with wearer preventing uncomfortable rubbing or chafing enhancing comfort and mobility
  • State-of-the-art 3-D mesh lining wicks moisture with one-handed quick connect buckles
  • Built-in belt loops
  • Torso buckles adjust easily, holding straps firmly in place thus completing five-point adjustment system
  • Soft, lightweight polyester webbing
  • Corrosion-resistant coated hardware
  • Washable harness with removable padding
  • Information and inspection logs are readily available in form of book-style labels
  • Integral lanyard keeper prevents tripping
  • Built-in impact load indicator gives notice about the fall of harness
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and the stringent ANSI Z359.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Style Vest