Learn More
3M™ DBI-Sala™ Salalift II Winch and Tripod Rescue Systems
Catalog No. 19809579 Shop All 3M Products
Change view
Click to view available options
Includes:
7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Galvanized Steel cable
7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Stainless Steel cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-809-579 7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Stainless Steel cable
19-066-856 7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Galvanized Steel cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-809-579 Supplier 3M™ Supplier No. 7100240697
With overload clutch that reduces chance of injury to entangled worker
  • Durable and lightweight corrosion resistant construction
  • With anodized aluminum side plates, zinc plated gears and shafts, stainless steel springs and aluminum drum
  • UL classified 7 in. (17.8cm) aluminum tripod
  • Quick mount bracket for winch attachment
  • Working load rating: 350 lb. (158.8kg) for work support and 310 lb. (140.6kg) for fall arrest
  • For raising, lowering, work support or personnel rescue
  • Comes with winch bag
  • With choice of 60 ft. (18.3m) of 0.25 in. (0.6cm) galvanized or stainless-steel cables
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL
For Use With (Application) Ideal for raising, lowering, work support or personnel rescue
Includes 7 ft. Aluminum tripod, winch salafit II, quick mount bracket, 60 ft. L of 0.25 in. dia. Stainless Steel cable