Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-Sala™ Suspension Trauma Safety Strap

Catalog No. 19809956 Shop All 3M Products
Change view
Click to view available options
Material:
Nylon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-809-956 Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-809-956 Supplier 3M™ Supplier No. 7100209339
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Helps to prevent the effects of suspension trauma after a fall

  • Allows worker who is suspended to stand up in their harness to relieve pressure
  • Safety accessory that can be attached to most harness brands/styles right in the field
  • Installation to a harness is fast and efficient — just choke off to strap and it's ready
  • Compact and lightweight design stays out of the worker's way
  • Easy deployment and operation: Simply unzip and hook straps together
  • Continuous-loop design allows for either one- or two-foot suspension relief without pinching
TRUSTED_SUSTAINABILITY

Specifications

Material Nylon
Weight (English) 3.2 oz.
Weight (Metric) 90.7g
Product Type Safety Strap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.