Learn More
3M™ Delta™ Vest Style Full Body Harnesses
Catalog No. 19056990 Shop All 3M Products
Change view
Click to view available options
D-Ring Location:
Back
Catalog No. D-Ring Location
19-056990 Back
1 options
Catalog No. 19-056990 Supplier 3M™ Supplier No. 7100231999
Add to Cart
Add to Cart
With back D-ring and tongue buckle legs
  • Spring-loaded, stand-up dorsal D-ring helps to make fast, easy and safe connections
  • i-Safe™system gives efficient, digital way of recording and storing information on inspections
  • Triangular design gives harness shape of its own
  • Quick and easy donning
  • Revolver™ torso adjusters makes adjustment fast and secure
  • Tech-Lite™ aluminum alloy buckles reduce carrying weight and energy required to do job
  • With basic shape, shoulder straps don't rub the neck raw
  • Soft, breathable material construction, hip pad ergonomically supports the back and hips while bearing heavy loads
  • Workhorse standing of 420 lb. carrying capacity
TRUSTED_SUSTAINABILITY
Specifications
Style Vest
D-Ring Location Back
Size X-Large