Learn More
3M™ DBI-Sala™ ExoFit NEX™ Construction Style Full Body Harness
Catalog No. 19809965 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-809-965 Large
19-809-966 X-Large
19-809-371 2X-Large
19-809-964 Medium
19-809-963 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-809-965 Supplier 3M™ Supplier No. 7100188763
Add to Cart
Add to Cart
Made for general construction work and have excellent tool-carrying capability
ExoFit NEX Construction Style Harness with aluminum front, back and side D-rings, locking quick connect buckles, sewn-in hip pad and removable belt.
  • Wraps around for the ultimate in no-tangle donning and comfortable security
  • Shoulder, hip and leg padding is built-in so it can't slip
  • Breathable lining guarantees stay-dry and comfortable every day
  • Memory-fit features allowing user to lock the adjustments in place to prevent the web from slipping
Compliance
Meets OSHA 1910.66 and OSHA 1926.502; ANSI™ A10.32, ANSI Z359.1, ANSI Z359.3 and ANSI Z359.4 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
D-Ring Location Back, Front, Side
Size Large
Style Construction Style