Learn More
Description
Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.
- For 10 and 12 mL serum transport tubes
Specifications
Specifications
| Diameter (Metric) Outer | 15 mm |
| Product Type | Screw Cap |
| Material | Polyethylene |
| Color | Red |
| Height (Metric) | 96 mm |
| No. per Case | 2000/Cs. |
| For Use With (Equipment) | Capitol Vial 10 and 12 mL Transport Tubes (not included) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
