Learn More
Bioventures Inc CAPPWash 8-Channel ELISA Plate Washer Head
Supplier:  Bioventures Inc W8
Encompass
CappWash is the ideal tool for low scale and reliable ELISA plate washing. Simple to use and economical, users can choose to simply order the manifold or the complete set, which includes a vacuum pump, bootles and tubing. Easy to operate Hold the manifold towards the plate and press the button to dispense the washing solution. Release the button to aspirate the liquid, which will go directly to the waste collection bottle Simple & robust design The CappWash is made not to fail.
Catalog No. NC1819161