Learn More
Kingfisher Biotech Inc Caprine GM-CSF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1093G005
Encompass
The Caprine GM-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Caprine GM-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Caprine GM-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Caprine GM-CSF Specifications: (Molecular Weight: 14.5 kDa) (Amino Acid Sequence: APTRQPSPVT WPWQHVDAIK EALSLLNDSS DTAAVMNETV EVVSEMFDSQ EPTCLQTRLE LYKQGLRGSL TSLTGSLTMM ASHYKKYCPP TQETSCETQI ITFKSFKENL KDFLFIIPFD CWEPVQK (127)) (Gene ID: 100861208). For research use only. Made in the USA
Catalog No. 50-253-3685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.