Learn More
Kingfisher Biotech Inc Caprine IFN alpha Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1177G100
Encompass_Preferred
The Caprine IFN alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Caprine IFN alpha applications are for cell culture, ELISA standard, and Western Blot Control. Caprine IFN alpha yeast-derived recombinant protein can be purchased in multiple sizes. Caprine IFN alpha Specifications: (Molecular Weight: 19.4 kDa) (Amino Acid Sequence: ICSLGCHLPH IHSLANGRVL MLLRQLRRVS PSSCLQDRKD FLFPQEALGG SQLQKAQALS VLHEVTQHTF QLFSTEGTAA AWDQSLLDKL RAALDQQLTD LQACLRQEEG LRGAPLLKED SSLAVRKYFH RVTLYLQEKG HSPCAWEVVR AEVMRAFSSS TNLQERFRRK D (171)) (Gene ID: 100860850). For research use only. Made in the USA
Catalog No. 50-253-3598
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.