Learn More
Kingfisher Biotech Inc Caprine IL-1 alpha Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1125G100
Encompass_Preferred
The Caprine IL-1 alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Caprine IL-1 alpha applications are for cell culture, ELISA standard, and Western Blot Control. Caprine IL-1 alpha yeast-derived recombinant protein can be purchased in multiple sizes. Caprine IL-1 alpha Specifications: (Molecular Weight: 17.9 kDa) (Amino Acid Sequence: SAHYSFQSNV KYNFMRVIHQ ECILNDALNQ SIIRDMSGPY LMAATLNNLE EAVKFDMVAY VSEEDSQLPV TLRISKTQLF VSAQNEDEPV LLKEMPETPK IIKDETNLLF FWEKHGSMDY FKSVAHPKLF IATKQEKLVH MASGPPSITD FQILEK (156)) (Gene ID: 100861289). For research use only. Made in the USA
Catalog No. 50-253-3648
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.