Learn More
Kingfisher Biotech Inc Caprine OPN Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1544G100
Encompass_Preferred
The Caprine Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Caprine Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Caprine Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Caprine Osteopontin Specifications: (Molecular Weight: 29.4 kDa) (Amino Acid Sequence: LPVKPTSSGS SEEKLLNNKY PDAVATWLKP DPSQKQTFLE PQNSVSSEET DDNKQNTLPS KSNESPEQTD DLDDDDETRH EVNSNDSDDA KTPDDSDHSN ESHHSDESDE ADFPTDIPTI AVFTPPFPTE STNDGRGDSV AYGLKSKSKK FRRSNVESPD ATEEDFTSHI ESEEMHDAPK KTSQLTDHSE ETNSDELSKE LTPKAQEESK HSNRIESQEN SKLSQEFHSL EDKLDLDHKS EEDKRLKIRI SHELDSASSK VN (262)) (Gene ID: n/a). For research use only. Made in the USA
Catalog No. 50-253-3170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.