Learn More
Research Products International Corp CAPS, 250 G
SDP
Supplier:  Research Products International Corp C30060250.0
Encompass_Preferred
pKa (25oC): 10.4. The working pH range in aqueous solution is 9.7-11.1
Catalog No. 50-197-7979
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.