Learn More
Biotang Inc CAPS, 500 GM, P-756, CAS # 1135-40-6. Purity > 99%, Soluble in water. Shipped at room temperature. Store at room temperature. Useful pH 9.7-11.1, mol. wt. 235.3.
Supplier:  Biotang Inc BTBB889
Encompass_Preferred
CAPS, 500 GM, P-756, CAS # 1135-40-6. Purity > 99%, Soluble in water. Shipped at room temperature. Store at room temperature. Useful pH 9.7-11.1, mol. wt. 235.3.
Catalog No. 50-152-343
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.