Learn More
Fisherbrand™ Caps for Auto-Analyzer Sample Cups
Catalog No. 22170180 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000/Cs.
Catalog No. Quantity
22-170-180 1000/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 22-170-180
Add to Cart
Add to Cart
This item is not returnable. View return policy
Produced from polyethylene (PE), these caps are designed specifically for use on our popular line of multi-purpose sample cups.
  • The caps are available with a solid or cross-cut pierceable top.
  • All caps snap securely in place on each of the sample cups.
Specifications
Product Type Pierceable Stopper
Material Polyethylene
Quantity 1000/Cs.
Sterility Non-sterile
For Use With (Equipment) Fisherbrand Auto-Analyzer Cups
Disposable Single-use