Learn More
Description
- Globe Scientific caps for automated systems fit several of the most popular platforms and fit seamlessly into capping processes
- These cost-effective alternatives to OEM caps are manufactured to the highest standards to ensure efficient sample processing
- Seamless cost-effective replacements for OEM caps
- Smooth operation in existing platforms
- Reliable sealing during high-speed transport and agitation
- Made from high-grade Polyethylene (PE) material
- Non-Sterile
Specifications
Specifications
| Product Type | Plug Cap |
| Closure Size | 13 mm |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Blue |
| Sterility | Non-sterile |
| For Use With (Equipment) | Beckman IDS Auto Track |
| Disposable | Single-use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.