Learn More
Sigma Aldrich Fine Chemicals Biosciences Tissue processing/embeddin
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z672122500EA
Disposable plastic cassettes hold tissue specimens during the embedding process as well as in a storage file. Molded from a special high density polymer these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.