Learn More
Microsolv Technology Corporation Caps, Headspace, Screw Top, Closed, with White Silicone Rubber / Red PTFE Liner, fitted in Steel, Magnetic, Silver Caps. For use with 18mm Precision Thread Headspace Vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9502045SCD
Encompass_Preferred
Silicone Rubber / Ultraclean PTFE Liner has a durometer of 45⁰ Shore A (hardness) and is 1.3mm thick. Cap has ND18 Screw Threads. Recommended with Agilent, Perkin Elmer Turbomax 16, 40 & 110 and other autosamplers. Closed Cap and liner good storage.
Catalog No. 50-227-1192
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.