Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPS, MP Biomedicals

Catalog No. ICN10143525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25g
100g
1kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10143525 25g
ICN10143580 100g
ICN10143591 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10143525 Supplier MP Biomedicals, Inc Supplier No. 0210143525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1135-40-6

Specifications

Quantity 25g
Chemical Name or Material CAPS
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.