Learn More
Microsolv Technology Corporation Caps, Push Top, with thinned penetration point, inserted in smooth, polyethylene, Blue Caps. For use with 8mm, Crimp Top autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9508C10CB
Encompass_Preferred
This cap is useful for non volatile solvents or when evaporation is not considered a problem.
Catalog No. 50-227-1266
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.