Learn More
Globe Scientific Caps for Sample Cups
Description
- For use on Globe Scientific sample cups
- Snap easily and securely into place
- Available with flat top (21-200-100) or pierceable cross cut (21-200-101)
- Polyethylene (PE)
- Natural color
Specifications
Specifications
| Closure Type | Flat Top |
| Quantity | 1000/Pk. |
| Color | Natural |
| Product Type | Snap Cap |
| Material | Polyethylene |
| For Use With (Equipment) | Globe Scientific Sample Cups |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.