Learn More
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | ≥99.0% |
| Solubility Information | Easily soluble in cold water |
| Formula Weight | 221.3 |
| Chemical Name or Material | zwitterionic buffer |
| Grade | by titration |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F). |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.