Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520517 Supplier Invitrogen™ Supplier No. PA520517
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20517 can be used with blocking peptide PEP-0637.

This gene encodes a novel neural/endocrine-specific cytosolic and peripheral membrane protein required for the Ca2+-regulated exocytosis of secretory vesicles. The protein acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4,5-bisphosphate P2). Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPS1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CADPS
Gene Accession No. Q62717, Q80TJ1, Q9ULU8
Gene Alias AU067781; Ca++-dependent secretion activator; Ca>2+>dependent activator protein for secretion; Ca>2+-dependent activator protein for secretion; Ca2+ dependent secretion activator; Ca2+-dependent activator protein; Ca2+-dependent activator protein for secretion; Ca2+-dependent secretion activator; Ca2+-regulated cytoskeletal protein; Cadps; CADPS1; calcium dependent secretion activator; calcium-dependent activator protein for secretion 1; Calcium-dependent secretion activator 1; Caps; Caps1; CAPS-1; Caps2; KIAA1121; mKIAA1121; rCAPS; UNC-31
Gene Symbols CADPS
Host Species Rabbit
Immunogen A 21 amino acid peptide near the amino terminus of the human CAPS1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26989, 27062, 8618
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.