Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CADPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103684
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103684 100 μL
1 options

Catalog No. PIPA5103684

Supplier: Thermo Scientific PA5103684

Only null left
Add to Cart
Add to Cart

CADPS2 Polyclonal Antibody

CADPS2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CADPS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CADPS2
Gene Accession No. Q86UW7
Gene Alias A230044C21Rik; Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Cadps2; calcium dependent secretion activator 2; calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; calcyphosin-2; calcyphosine 2; Calcyphosine-2; calcyphosphine 2; Caps2; CAPS-2; cerebellum postnatal development associated protein 2; Cpd2; KIAA1591; RGD1559440; UG0636c06
Gene Symbols CADPS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CADPS2(Accession Q86UW7), corresponding to amino acid residues A48-P98.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312166, 93664
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.