Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018501 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8501 50 mg
11-101-9937 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8501 Supplier MilliporeSigma Supelco Supplier No. 1208450MGF
Add to Cart
Add to Cart

Superior quality of pure E-Capsaicin, with a guaranteed assay of =99.0% by HPLC.

Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Description Analytical standard for food analysis
CAS 404-86-4
Flash Point 113°C
Physical Form Neat
Linear Formula (CH3)2 CHCH=CH(CH2)4 CONHCH2C6H3-4-(OH)-3-(OCH3)
MDL Number MFCD00017259
Melting Point 62°C to 65°C
Synonym 8-Methyl-N-vanillyl-trans-6-nonenamide
Solubility Insoluble in water
Type Analytical Standard
Quantity 50 mg
Formula Weight 305.41
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.