Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ CapSeal Bullet™ Capillary Ferrule, General Purpose, 0.32 mm Column I.D.

Catalog No. 111010715 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
12/Pk.
48/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-101-0715 12/Pk.
11-100-9095 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-0715 Supplier MilliporeSigma™ Supelco™ Supplier No. 23481
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capillary ferrules have a precision bore which is centrally located. During assembly of a connector, fiber is fixed within the ferrule using adhesive before grinding and polishing for end preparation.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 0.5 mm
Packaging Quantity 12/Pk.
Description Ferrule Type: General Purpose

CapSeal Bullet is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.