Learn More
ACROBiosystems HIV (HV1-HXB2) Capsid protein p24, His Tag (MALS verified)
Supplier: ACROBiosystems CP4H52H3100UG
            HIV (HV1-HXB2) Capsid protein p24, His Tag (CP4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Pro 133 - Leu 363 (Accession # P04591).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.