Learn More
Cytiva 70K Minimate™ Tangential Flow Filtration (TFF) capsule with Omega™ membrane (1/pkg)
Supplier:  Cytiva OA070C12
Encompass_Preferred
Reusable TFF Device for Ultrafiltration Applications. Accelerates and Simplifies Processing of Up to 1 L-Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system.-Achieves high concentration factors in a single step due to the low holdup volume of the device, as little as 5 mL.-Capsule includes all fittings and tubing to simplify installation into filtration setups.-Several Minimate TFF capsules may be connected in parallel for increased membrane area. -Capsules have the same path length and materials of construction as larger Pall Centramate™ systems used in process development and production.-Optimized flow path design coupled with low protein binding Omega membrane minimizes non-specific binding.-100% integrity tested during manufacture, for critical applications, the user can re-test the integrity after initial use. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9566
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.